4-Brom-2-fluor-6-nitroanilin
Lieferant: ACTIVATE SCIENTIFIC
AS33835G1
AS33835G10
AS33835G5
ACTIAS33835G1EA
0
CHF
ACTIAS33835G1
ACTIAS33835G10
ACTIAS33835G5
4-Brom-2-fluor-6-nitroanilin
4-Brom-2-fluor-6-nitroanilin
Formel:
C₆H₄BrFN₂O₂ |
MDL Number:
MFCD03094271 CAS-Nummer: 517920-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...