2-Brom-4-fluor-6-nitroanilin

Lieferant: ACTIVATE SCIENTIFIC

AS35169G1 AS35169G10 AS35169G5
ACTIAS35169G1EA 0 CHF
ACTIAS35169G1 ACTIAS35169G10 ACTIAS35169G5
2-Brom-4-fluor-6-nitroanilin
2-Brom-4-fluor-6-nitroanilin
Formel: C₆H₄BrFN₂O₂
MDL Number: MFCD07779528
CAS-Nummer: 10472-88-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR