3-(N,N-Dimethylamino)phenylboronsäure Hydrochlorid

Lieferant: ACTIVATE SCIENTIFIC

AS33724G1 AS33724G5
ACTIAS33724G1EA 0 CHF
ACTIAS33724G1 ACTIAS33724G5
3-(N,N-Dimethylamino)phenylboronsäure Hydrochlorid
3-(N,N-Dimethylamino)phenylboronsäure Hydrochlorid
Formel: C₈H₁₃BClNO₂
MW: 201,4583 g/mol
MDL Number: MFCD04112542
CAS-Nummer: 1256355-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR