3-(N,N-Dimethylamino)phenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Dimethylaminophenylboronic acid, 3-(Dimethylamino)benzeneboronic acid

AS50339G1 AS50339G5
ACTIAS50339G1EA 0 CHF
ACTIAS50339G1 ACTIAS50339G5
3-(N,N-Dimethylamino)phenylboronsäure
3-(N,N-Dimethylamino)phenylboronsäure
Formel: C₈H₁₂BNO₂
MDL Number: MFCD01318996
CAS-Nummer: 178752-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR