3-Carboxy-5-nitrophenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
AS50061G1
AS50061G5
ACTIAS50061G1EA
0
CHF
ACTIAS50061G1
ACTIAS50061G5
3-Carboxy-5-nitrophenylboronsäure
3-Carboxy-5-nitrophenylboronsäure
Formel:
C₇H₆BNO₆ |
MDL Number:
MFCD00757433 CAS-Nummer: 101084-81-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...