4-Carboxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Boronobenzoic Acid, Ark Pharm, 4-Carboxybenzeneboronic acid

AS50109G100 AS50109G5
ACTIAS50109G100EA 0 CHF
ACTIAS50109G100 ACTIAS50109G5
4-Carboxyphenylboronsäure
4-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
MW: 165,94 g/mol
MDL Number: MFCD00151801
CAS-Nummer: 14047-29-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR