3-Carboxyphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Boronobenzoic acid, 3-Carboxybenzeneboronic acid

AS50097G100 AS50097G5
ACTIAS50097G100EA 0 CHF
ACTIAS50097G100 ACTIAS50097G5
3-Carboxyphenylboronsäure
3-Carboxyphenylboronsäure
Formel: C₇H₇BO₄
MW: 165,94 g/mol
MDL Number: MFCD00036833
CAS-Nummer: 25487-66-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR