2-(2-Chlorethoxy)-4'-nitroacetanilid
Lieferant: ACTIVATE SCIENTIFIC
AS23714G1
AS23714G5
ACTIAS23714G1EA
0
CHF
ACTIAS23714G1
ACTIAS23714G5
2-(2-Chlorethoxy)-4'-nitroacetanilid
2-(2-Chlorethoxy)-4'-nitroacetanilid
Formel:
C₁₀H₁₁ClN₂O₄ |
MDL Number:
MFCD18379274 CAS-Nummer: 811450-82-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...