1-(Phenylsulfonyl)-2-indolboronsäure

Lieferant: ACTIVATE SCIENTIFIC

AS51499G1 AS51499G5
ACTIAS51499G1EA 0 CHF
ACTIAS51499G1 ACTIAS51499G5
1-(Phenylsulfonyl)-2-indolboronsäure
1-(Phenylsulphonyl)-2-indoleboronic acid
Formel: C₁₄H₁₂BNO₄S
MDL Number: MFCD03086094
CAS-Nummer: 342404-46-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR