5-Indolboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
(1H-indol-5-yl)boronic acid, (1H-Indol-5-yl)boronic acid, 5-Indolylboronic acid
AS50837G1
AS50837G5
ACTIAS50837G1EA
0
CHF
ACTIAS50837G1
ACTIAS50837G5
5-Indolboronsäure
5-Indoleboronic acid
Formel:
C₈H₈BNO₂ MW: 160,97 g/mol |
MDL Number:
MFCD01319013 CAS-Nummer: 144104-59-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...