4-Indolboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Indolylboronic acid, (1H-Indol-4-yl)boronic acid
I2051G5
AS50041G1
AS50041G5
XXXXI2051G5EA
0
CHF
XXXXI2051G5
ACTIAS50041G1
ACTIAS50041G5
4-Indolboronsäure
4-Indoleboronic acid
Formel:
C₈H₈BNO₂ MW: 160,97 g/mol |
MDL Number:
MFCD03095175 CAS-Nummer: 220465-43-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...