2,5-Furandicarbonsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
2,5-Furandicarboxylic acid
AS32768G1
AS32768G10
AS32768G5
ACTIAS32768G1EA
0
CHF
ACTIAS32768G1
ACTIAS32768G10
ACTIAS32768G5
2,5-Furandicarbonsäure
Furan-2,5-dicarboxylic acid
Formel:
C₆H₄O₅ MW: 156,09 g/mol |
MDL Number:
MFCD00016582 CAS-Nummer: 3238-40-2 EINECS: 221-800-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...