4-Chlor-3-methylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-3-methylbenzeneboronic acid

AS35292G1 AS35292G5
ACTIAS35292G1EA 0 CHF
ACTIAS35292G1 ACTIAS35292G5
4-Chlor-3-methylphenylboronsäure
4-Chlor-3-methylphenylboronsäure
Formel: C₇H₈BClO₂
MW: 170,40 g/mol
MDL Number: MFCD02683104
CAS-Nummer: 161950-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR