4-Chlor-2-methylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Chloro-2-methylbenzeneboronic acid

AS31791G1 AS31791G5
ACTIAS31791G1EA 0 CHF
ACTIAS31791G1 ACTIAS31791G5
4-Chlor-2-methylphenylboronsäure
4-Chlor-2-methylphenylboronsäure
Formel: C₇H₈BClO₂
MW: 170,40 g/mol
MDL Number: MFCD02683107
CAS-Nummer: 209919-30-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR