3-Chlor-4-methylphenylboronsäure
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Chloro-4-methylphenylboronic Acid, 3-Chloro-4-methylbenzeneboronic acid
AS34968G1
AS34968G10
AS34968G5
ACTIAS34968G1EA
0
CHF
ACTIAS34968G1
ACTIAS34968G10
ACTIAS34968G5
3-Chlor-4-methylphenylboronsäure
3-Chlor-4-methylphenylboronsäure
Formel:
C₇H₈BClO₂ MW: 170,40 g/mol |
MDL Number:
MFCD04039010 CAS-Nummer: 175883-63-3 |