6-Hydroxy-5-nitronicotinsäure
Lieferant: ACTIVATE SCIENTIFIC
AS22423G25
AS22423G5
ACTIAS22423G25EA
110
CHF
ACTIAS22423G25
ACTIAS22423G5
6-Hydroxy-5-nitronicotinsäure
6-Hydroxy-5-nitronicotinsäure
Formel:
C₆H₄N₂O₅ MW: 184,1100 g/mol |
MDL Number:
MFCD02180840 CAS-Nummer: 6635-31-0 |
Learn more
![](/m-ch.vwr.com/de_CH/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...