3-Methyl-1H-1,2,4-triazol

Lieferant: ACTIVATE SCIENTIFIC

AS36453G1 AS36453G5
ACTIAS36453G1EA 0 CHF
ACTIAS36453G1 ACTIAS36453G5
3-Methyl-1H-1,2,4-triazol
3-Methyl-1H-1,2,4-triazol
Formel: C₃H₅N₃
MW: 83,0900 g/mol
MDL Number: MFCD00233931
CAS-Nummer: 7170-01-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR