5-Pyrimidincarbonitril

Lieferant: ACTIVATE SCIENTIFIC

AS30400G1 AS30400G5
ACTIAS30400G1EA 0 CHF
ACTIAS30400G1 ACTIAS30400G5
5-Pyrimidincarbonitril
5-Pyrimidincarbonitril
Formel: C₅H₃N₃
MW: 105,1 g/mol
MDL Number: MFCD00235052
CAS-Nummer: 40805-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR