2,6-Dibrom-9,10-anthrachinon

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2,6-Dibromoanthracene-9,10-dione

AS32791G1 AS32791G5
ACTIAS32791G1EA 0 CHF
ACTIAS32791G1 ACTIAS32791G5
2,6-Dibrom-9,10-anthrachinon
2,6-Dibrom-9,10-anthrachinon
Formel: C₁₄H₆Br₂O₂
MDL Number: MFCD08276344
CAS-Nummer: 633-70-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR