2-Brom-6-fluornaphthalin
Lieferant: ACTIVATE SCIENTIFIC
AS35852G1
AS35852G5
ACTIAS35852G1EA
103
CHF
ACTIAS35852G1
ACTIAS35852G5
2-Brom-6-fluornaphthalin
2-Bromo-6-fluoronaphthalene
Formel:
C₁₀H₆BrF MW: 225,0600 g/mol |
MDL Number:
MFCD09763683 CAS-Nummer: 324-41-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...