Ethyl-4-brompicolinat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Ethyl 4-bromopicolinate

AS35821G1 AS35821G5
ACTIAS35821G1EA 0 CHF
ACTIAS35821G1 ACTIAS35821G5
Ethyl-4-brompicolinat
Ethyl-4-brompicolinat
Formel: C₈H₈BrNO₂
MW: 230,0600 g/mol
MDL Number: MFCD09839150
CAS-Nummer: 62150-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR