Methyl-4-fluorsalicylat
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
Methyl 4-fluoro-2-hydroxybenzoate
AS33279G1
AS33279G5
ACTIAS33279G1EA
0
CHF
ACTIAS33279G1
ACTIAS33279G5
Methyl-4-fluorsalicylat
Methyl-4-fluorsalicylat
Formel:
C₈H₇FO₃ |
MDL Number:
MFCD02093965 CAS-Nummer: 392-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...