Methyl-4-brom-3-hydroxybenzoat

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: Methyl 4-bromo-3-hydroxybenzoate

AS34308G1 AS34308G5
ACTIAS34308G1EA 0 CHF
ACTIAS34308G1 ACTIAS34308G5
Methyl-4-brom-3-hydroxybenzoat
Methyl-4-brom-3-hydroxybenzoat
Formel: C₈H₇BrO₃
MW: 231,0400 g/mol
MDL Number: MFCD08056372
CAS-Nummer: 106291-80-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR