1-Brom-3,5-di-tert-butylbenzol

Lieferant: ACTIVATE SCIENTIFIC

AS33083G1 AS33083G5
ACTIAS33083G1EA 0 CHF
ACTIAS33083G1 ACTIAS33083G5
1-Brom-3,5-di-tert-butylbenzol
1-Brom-3,5-di-tert-butylbenzol
Formel: C₁₄H₂₁Br
MDL Number: MFCD00796945
CAS-Nummer: 22385-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR