3-Brom-5-methylanisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromo-5-methoxytoluene, 1-Bromo-3-methoxy-5-methylbenzene

AS34977G1 AS34977G10 AS34977G5
ACTIAS34977G1EA 0 CHF
ACTIAS34977G1 ACTIAS34977G10 ACTIAS34977G5
3-Brom-5-methylanisol
3-Brom-5-methylanisol
Formel: C₈H₉BrO
MW: 201,0600 g/mol
MDL Number: MFCD08061916
CAS-Nummer: 29578-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR