3-Fluor-2-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS33670G1 AS33670G10 AS33670G5
ACTIAS33670G1EA 0 CHF
ACTIAS33670G1 ACTIAS33670G10 ACTIAS33670G5
3-Fluor-2-nitrobenzoesäure
3-Fluoro-2-nitrobenzoic acid
Formel: C₇H₄FNO₄
MW: 185.11 g/mol
MDL Number: MFCD07368342
CAS-Nummer: 1000339-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR