2-(Chlormethyl)benzothiazol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-(Chloromethyl)-1,3-benzothiazole

AS35999G1 AS35999G5
ACTIAS35999G1EA 0 CHF
ACTIAS35999G1 ACTIAS35999G5
2-(Chlormethyl)benzothiazol
2-(Chloromethyl)benzothiazole
Formel: C₈H₆CINS
MW: 183,65 g/mol
MDL Number: MFCD05664944
CAS-Nummer: 37859-43-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR