2-Chlor-4-fluor-5-nitrobenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS32646G1 AS32646G5
ACTIAS32646G1EA 0 CHF
ACTIAS32646G1 ACTIAS32646G5
2-Chlor-4-fluor-5-nitrobenzoesäure
2-Chloro-4-fluoro-5-nitrobenzoic acid
Formel: C₇H₃ClFNO
MW: 219,55 g/mol
MDL Number: MFCD04115715
CAS-Nummer: 114776-15-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR