2,6-Dichlor-3-fluorbenzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS35514G1 AS35514G5
ACTIAS35514G1EA 0 CHF
ACTIAS35514G1 ACTIAS35514G5
2,6-Dichlor-3-fluorbenzoesäure
2,6-Dichloro-3-fluorobenzoic acid
Formel: C₇H₃Cl₂FO₂
MW: 209,0019 g/mol
MDL Number: MFCD06658165
CAS-Nummer: 178813-78-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR