2,4-Dichlor-3-fluorbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32144G1 AS32144G5
ACTIAS32144G1EA 0 CHF
ACTIAS32144G1 ACTIAS32144G5
2,4-Dichlor-3-fluorbenzonitril
2,4-Dichlor-3-fluorbenzonitril
Formel: C₇H₂Cl₂FN
MW: 190,0000 g/mol
MDL Number: MFCD06658268
CAS-Nummer: 161612-68-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR