3-Brom-5-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 1-Bromo-3-methoxy-5-nitrobenzene

AS35520G1 AS35520G5
ACTIAS35520G1EA 0 CHF
ACTIAS35520G1 ACTIAS35520G5
3-Brom-5-nitroanisol
3-Brom-5-nitroanisol
Formel: C₇H₆BrNO₃
MW: 232,0300 g/mol
MDL Number: MFCD04112752
CAS-Nummer: 16618-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR