(4-Cyanphenyl)essigsäuremethylester
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
4-Cyanobenzeneacetic acid methyl ester, Methyl (4-cyanophenyl)acetate
AS36009G1
AS36009G5
ACTIAS36009G1EA
0
CHF
ACTIAS36009G1
ACTIAS36009G5
(4-Cyanphenyl)essigsäuremethylester
(4-Cyanphenyl)essigsäuremethylester
Formel:
C₁₀H₉NO₂ |
MDL Number:
MFCD02181140 CAS-Nummer: 52798-01-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...