3-Fluor-5-iodbenzonitril

Lieferant: ACTIVATE SCIENTIFIC

AS32462G1 AS32462G5
ACTIAS32462G1EA 0 CHF
ACTIAS32462G1 ACTIAS32462G5
3-Fluor-5-iodbenzonitril
3-Fluor-5-iodbenzonitril
Formel: C₇H₃FIN
MW: 247,0100 g/mol
MDL Number: MFCD04116468
CAS-Nummer: 723294-75-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR