2-Cyan-6-fluoranilin

Lieferant: ACTIVATE SCIENTIFIC

AS32235G1 AS32235G10 AS32235G5
ACTIAS32235G1EA 0 CHF
ACTIAS32235G1 ACTIAS32235G10 ACTIAS32235G5
2-Cyan-6-fluoranilin
2-Cyan-6-fluoranilin
Formel: C₇H₅FN₂
MDL Number: MFCD07368774
CAS-Nummer: 115661-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR