3-Fluorsalicylaldehyd
Lieferant: ACTIVATE SCIENTIFIC
Synonyms:
3-Fluoro-2-hydroxybenzaldehyde
AS30964G10
AS30964G5
ACTIAS30964G10EA
0
CHF
ACTIAS30964G10
ACTIAS30964G5
3-Fluorsalicylaldehyd
3-Fluorsalicylaldehyd
Formel:
C₇H₅FO₂ |
MDL Number:
MFCD00003319 CAS-Nummer: 394-50-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...