2-Amino-4,6-dibrompyrimidin

Lieferant: ACTIVATE SCIENTIFIC

AS30535G1 AS30535G5
ACTIAS30535G1EA 0 CHF
ACTIAS30535G1 ACTIAS30535G5
2-Amino-4,6-dibrompyrimidin
2-Amino-4,6-dibromopyrimidine
Formel: C₄H₃Br₂N₃
MDL Number: MFCD09263251
CAS-Nummer: 856973-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR