3,6-Difluorpicolinsäure
Lieferant: ACTIVATE SCIENTIFIC
AS51438G1
AS51438G5
ACTIAS51438G1EA
0
CHF
ACTIAS51438G1
ACTIAS51438G5
3,6-Difluorpicolinsäure
3,6-Difluorpicolinsäure
Formel:
C₆H₃F₂NO₂ MW: 159,0900 g/mol |
MDL Number:
MFCD09031176 CAS-Nummer: 851386-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...