4-Amino-α,α,α-trifluor-o-toluylsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 4-Amino-2-(trifluoromethyl)benzoic acid

AS22660G100 AS22660G25
ACTIAS22660G100EA 389 CHF
ACTIAS22660G100 ACTIAS22660G25
4-Amino-α,α,α-trifluor-o-toluylsäure
4-Amino-α,α,α-trifluor-o-toluylsäure
Formel: C₈H₆F₃NO₂
MW: 205,14 g/mol
MDL Number: MFCD03407959
CAS-Nummer: 393-06-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR