4-Brom-2-(trifluormethyl)benzoesäure
Lieferant: ACTIVATE SCIENTIFIC
AS31244G1
AS31244G5
ACTIAS31244G1EA
0
CHF
ACTIAS31244G1
ACTIAS31244G5
4-Brom-2-(trifluormethyl)benzoesäure
4-Bromo-2-(trifluoromethyl)benzoic acid
Formel:
C₈H₄BrF₃O₂ MW: 269.02 g/mol |
MDL Number:
MFCD07787468 CAS-Nummer: 320-31-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...