3-Brom-4-(trifluormethyl)benzoesäure

Lieferant: ACTIVATE SCIENTIFIC

AS36545G1 AS36545G5
ACTIAS36545G1EA 0 CHF
ACTIAS36545G1 ACTIAS36545G5
3-Brom-4-(trifluormethyl)benzoesäure
3-Bromo-4-(trifluoromethyl)benzoic acid
Formel: C₈H₄BrF₃O₂
MW: 269.0200 g/mol
MDL Number: MFCD13185386
CAS-Nummer: 581813-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR