3-Fluor-2-nitroanisol

Lieferant: ACTIVATE SCIENTIFIC

AS32444G1 AS32444G5
ACTIAS32444G1EA 0 CHF
ACTIAS32444G1 ACTIAS32444G5
3-Fluor-2-nitroanisol
3-Fluor-2-nitroanisol
Formel: C₇H₆FNO₃
MW: 171,1259 g/mol
MDL Number: MFCD07368752
CAS-Nummer: 641-49-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR