2-Fluor-m-toluidin

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-3-methylaniline, 3-Amino-2-fluorotoluene

AS33734G1 AS33734G10 AS33734G5
ACTIAS33734G1EA 0 CHF
ACTIAS33734G1 ACTIAS33734G10 ACTIAS33734G5
2-Fluor-m-toluidin
2-Fluor-m-toluidin
Formel: C₇H₈FN
MW: 125,14 g/mol
MDL Number: MFCD06410915
CAS-Nummer: 1978-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR