2-Fluor-4-methylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Fluoro-4-methylbenzeneboronic acid, (2-Fluoro-4-methylphenyl)boronic acid

AS31550G1 AS31550G5
ACTIAS31550G1EA 0 CHF
ACTIAS31550G1 ACTIAS31550G5
2-Fluor-4-methylphenylboronsäure
2-Fluor-4-methylphenylboronsäure
Formel: C₇H₈BFO₂
MW: 153,95 g/mol
MDL Number: MFCD05664239
CAS-Nummer: 170981-26-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR