4-Brom-3-(trifluormethyl)anisol

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Bromo-5-methoxybenzotrifluoride, 4-Bromo-3-trifluoromethylanisole

AS31277G1 AS31277G10 AS31277G5
ACTIAS31277G1EA 0 CHF
ACTIAS31277G1 ACTIAS31277G10 ACTIAS31277G5
4-Brom-3-(trifluormethyl)anisol
4-Brom-3-(trifluormethyl)anisol
Formel: C₈H₆BrF₃O
MW: 255,04 g/mol
MDL Number: MFCD08059527
CAS-Nummer: 400-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR