2-Isopropylphenylboronsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Isopropylbenzeneboronic acid, (2-Isopropylphenyl)boronic acid

AS35481G1 AS35481G5
ACTIAS35481G1EA 0 CHF
ACTIAS35481G1 ACTIAS35481G5
2-Isopropylphenylboronsäure
2-Isopropylphenylboronsäure
Formel: C₉H₁₃BO₂
MW: 164,01 g/mol
MDL Number: MFCD03411937
CAS-Nummer: 89787-12-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR