3-Methylsalicylaldehyd

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 2-Hydroxy-3-methylbenzaldehyde

AS35113G1 AS35113G10 AS35113G5
ACTIAS35113G1EA 0 CHF
ACTIAS35113G1 ACTIAS35113G10 ACTIAS35113G5
3-Methylsalicylaldehyd
3-Methylsalicylaldehyd
Formel: C₈H₈O₂
MDL Number: MFCD00040798
CAS-Nummer: 824-42-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR