2-Aminochinazolin

Lieferant: ACTIVATE SCIENTIFIC

AS30926G1 AS30926G5
ACTIAS30926G1EA 0 CHF
ACTIAS30926G1 ACTIAS30926G5
2-Aminochinazolin
2-Aminoquinazoline
Formel: C₈H₇N₃
MW: 145,1600 g/mol
MDL Number: MFCD00956186
CAS-Nummer: 1687-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR