4,7-Dibrom-2,1,3-benzothiadiazol

Lieferant: ACTIVATE SCIENTIFIC

AS31772G1 AS31772G5
ACTIAS31772G1EA 0 CHF
ACTIAS31772G1 ACTIAS31772G5
4,7-Dibrom-2,1,3-benzothiadiazol
4,7-Dibromo-2,1,3-benzothiadiazole
Formel: C₆H₂Br₂N₂S
MDL Number: MFCD00658844
CAS-Nummer: 15155-41-6
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR