2-Nitro-4-thiophencarbonsäure

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 5-Nitrothiophene-3-carboxylic acid

AS35956G1 AS35956G5
ACTIAS35956G1EA 0 CHF
ACTIAS35956G1 ACTIAS35956G5
2-Nitro-4-thiophencarbonsäure
2-Nitro-4-thiophencarbonsäure
Formel: C₅H₃NO₄S
MW: 173,15 g/mol
MDL Number: MFCD00052582
CAS-Nummer: 40357-96-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR