3-Brom-2-thiophencarbonitril

Lieferant: ACTIVATE SCIENTIFIC

Synonyms: 3-Bromothiophene-2-carbonitrile

AS1737G1 AS1737G5
ACTIAS1737G1EA 0 CHF
ACTIAS1737G1 ACTIAS1737G5
3-Brom-2-thiophencarbonitril
3-Brom-2-thiophencarbonitril
Formel: C₅H₂BrNS
MW: 188,05 g/mol
MDL Number: MFCD00068173
CAS-Nummer: 18791-98-5
UN: 3439
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR